missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG7 Rabbit anti-Human, Mouse, Rat, Clone: 3X1W4, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £397.00
Specifications
| Antigen | ATG7 |
|---|---|
| Clone | 3X1W4 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18387835
|
Novus Biologicals
NBP3-15808-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048455
|
Novus Biologicals
NBP3-15808-100UL |
100 μg |
£397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG7 Monoclonal antibody specifically detects ATG7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDownSpecifications
| ATG7 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Atg7(Apg7) (O95352). MAAATGDPGLSKLQFAPFSSALDVGFWHELTQKKLNEYRLDEAPKDIKGYYYNGDSAGLPARLTLEFSAFDMSAPTPARCCPAIGTLYNTNTLESFKTAD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 3X1W4 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Autophagy | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 10533 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title