missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4D Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £359.00
Specifications
| Antigen | ATG4D |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666432
|
Novus Biologicals
NBP2-92681-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696142
|
Novus Biologicals
NBP2-92681-0.1ml |
0.1 mL |
£359.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG4D Polyclonal antibody specifically detects ATG4D in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ATG4D | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy | |
| PBS with 50% glycerol, pH7.3. | |
| 84971 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG4 autophagy 4 homolog D, APG4 autophagy 4 homolog D (S. cerevisiae), APG4D, APG4-D, ATG4 autophagy related 4 homolog D (S. cerevisiae), AUTL4, AUT-like 4 cysteine endopeptidase, AUT-like 4, cysteine endopeptidase, AUT-like 4, cysteine endopeptidase (S. cerevisiae), Autophagin-4, Autophagy-related cysteine endopeptidase 4, Autophagy-related protein 4 homolog D, cysteine protease ATG4D, cysteine protease involved in autophagy, EC 3.4.22, EC 3.4.22.- | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-474 of human ATG4D (NP_116274.3). DVTRLVVYVSQDCTVYKADVARLVARPDPTAEWKSVVILVPVRLGGETLNPVYVPCVKELLRCELCLGIMGGKPRHSLYFIGYQDDFLLYLDPHYCQPTVDVSQADFPLESFHCTSPRKMAFAKMDPSCTVGFYAGDRKEFETLCSELTRVLSSSSATERYPMFTLAEGHAQDHSLDDLCSQLAQPTLRLPRTGRLLRAKRPSSEDFVFL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title