missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4D Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92681-0.1ml
This item is not returnable.
View return policy
Description
ATG4D Polyclonal antibody specifically detects ATG4D in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ATG4D | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| APG4 autophagy 4 homolog D, APG4 autophagy 4 homolog D (S. cerevisiae), APG4D, APG4-D, ATG4 autophagy related 4 homolog D (S. cerevisiae), AUTL4, AUT-like 4 cysteine endopeptidase, AUT-like 4, cysteine endopeptidase, AUT-like 4, cysteine endopeptidase (S. cerevisiae), Autophagin-4, Autophagy-related cysteine endopeptidase 4, Autophagy-related protein 4 homolog D, cysteine protease ATG4D, cysteine protease involved in autophagy, EC 3.4.22, EC 3.4.22.- | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-474 of human ATG4D (NP_116274.3). DVTRLVVYVSQDCTVYKADVARLVARPDPTAEWKSVVILVPVRLGGETLNPVYVPCVKELLRCELCLGIMGGKPRHSLYFIGYQDDFLLYLDPHYCQPTVDVSQADFPLESFHCTSPRKMAFAKMDPSCTVGFYAGDRKEFETLCSELTRVLSSSSATERYPMFTLAEGHAQDHSLDDLCSQLAQPTLRLPRTGRLLRAKRPSSEDFVFL | |
| 0.1 mL | |
| Autophagy | |
| 84971 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction