missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ATG16L2 |
|---|---|
| Concentration | 0.33 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16822805
|
Proteintech
24322-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16812805
|
Proteintech
24322-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in autophagy By similarity. Ota T., Nat. Genet. 36:40-45(2004). Mural R. J., Submitted (JUL-2005) to the EMBL/GenBank/DDBJ databases. Bechtel S., BMC Genomics 8:399-399(2007).Specifications
| ATG16L2 | |
| Immunofluorescence, Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q6KAU8, Q8NAA4 | |
| 73683, 89849 | |
| ATG16L2 Fusion Protein Ag19399 | |
| Primary | |
| -20°C | |
| ATG16L2 |
| 0.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| APG16 like 2, ATG16L2, Autophagy related protein 16 2, WDR80 | |
| Atg16l2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title