missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24322-1-AP-20UL
This item is not returnable.
View return policy
Description
May play a role in autophagy By similarity. Ota T., Nat. Genet. 36:40-45(2004). Mural R. J., Submitted (JUL-2005) to the EMBL/GenBank/DDBJ databases. Bechtel S., BMC Genomics 8:399-399(2007).Specifications
| ATG16L2 | |
| Polyclonal | |
| Unconjugated | |
| ATG16L2 | |
| APG16 like 2, ATG16L2, Autophagy related protein 16 2, WDR80 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 73683, 89849 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6KAU8, Q8NAA4 | |
| Atg16l2 | |
| ATG16L2 Fusion Protein Ag19399 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion