missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG13 Recombinant Rabbit Monoclonal Antibody (4L5P5)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | ATG13 |
|---|---|
| Clone | 4L5P5 |
| Concentration | 2.24 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284391
|
Invitrogen™
MA557915 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30284392
![]() |
Invitrogen™
MA557916 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: ADLAYPVVFA AGLNATHPHQ LMVPGKEGGV PLAPNQPVHG TQADQERLAT CTPSDRTHCA ATPSSSEDTE TVSNSSEGRA SPHDVLETIF VRKVGAFVNK PINQVTLTSL DIPFAMFAPK NLELEDTDPM VNPPDSPETE SPLQGSLHSD GSSGGSSGNT HDDFVMIDFK PAFSKDDILP MDLGTFYREF QNPPQLSSLS.
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein ATG1. ATG13 forms a complex with ULK1 and ULK2, the mammalian homologs of ATG1, and with FIP200. This complex is a target of TOR phosphorylation under normal conditions; inhibition of TOR by rapamycin or leucine deprivation leads to dephosphorylation of ATG13, ULK1 and ULK2, which then leads to autophagy. Knockdown of ATG13 inhibits autophagosome formation.Specifications
| ATG13 | |
| 2.24 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O75143, Q91YI1 | |
| 362164, 51897, 9776 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 281-480 of human ATG13 (NP_001136145.1). | |
| Antibody |
| 4L5P5 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ATG13 | |
| 1110053A20Rik; Atg13; ATG13 autophagy related 13 homolog; autophagy related 13; autophagy-related protein 13; D2Ertd391e; FLJ20698; Harbi1; Harbi1l; harbinger transposase derived 1; KIAA0652; OTTHUMP00000233321; OTTHUMP00000233322; OTTHUMP00000233323; OTTHUMP00000233324; OTTHUMP00000233325; OTTHUMP00000233326; PARATARG8; RGD1310685 | |
| ATG13 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title