missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATG13 Recombinant Rabbit Monoclonal Antibody (4L5P5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557915

 View more versions of this product

Product Code. 30284391

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: ADLAYPVVFA AGLNATHPHQ LMVPGKEGGV PLAPNQPVHG TQADQERLAT CTPSDRTHCA ATPSSSEDTE TVSNSSEGRA SPHDVLETIF VRKVGAFVNK PINQVTLTSL DIPFAMFAPK NLELEDTDPM VNPPDSPETE SPLQGSLHSD GSSGGSSGNT HDDFVMIDFK PAFSKDDILP MDLGTFYREF QNPPQLSSLS.

Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein ATG1. ATG13 forms a complex with ULK1 and ULK2, the mammalian homologs of ATG1, and with FIP200. This complex is a target of TOR phosphorylation under normal conditions; inhibition of TOR by rapamycin or leucine deprivation leads to dephosphorylation of ATG13, ULK1 and ULK2, which then leads to autophagy. Knockdown of ATG13 inhibits autophagosome formation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATG13
Recombinant Monoclonal
2.24 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
O75143, Q91YI1
ATG13
Recombinant fusion protein containing a sequence corresponding to amino acids 281-480 of human ATG13 (NP_001136145.1).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Western Blot
4L5P5
Unconjugated
ATG13
1110053A20Rik; Atg13; ATG13 autophagy related 13 homolog; autophagy related 13; autophagy-related protein 13; D2Ertd391e; FLJ20698; Harbi1; Harbi1l; harbinger transposase derived 1; KIAA0652; OTTHUMP00000233321; OTTHUMP00000233322; OTTHUMP00000233323; OTTHUMP00000233324; OTTHUMP00000233325; OTTHUMP00000233326; PARATARG8; RGD1310685
Rabbit
Protein A
RUO
362164, 51897, 9776
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.