missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG13 Antibody (6B2E12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | ATG13 |
|---|---|
| Clone | 6B2E12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18265820
|
Novus Biologicals
NBP2-52418 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659696
|
Novus Biologicals
NBP2-52418-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG13 Monoclonal specifically detects ATG13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ATG13 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| PBS with 0.05% Sodium Azide | |
| 9776 | |
| Purified recombinant fragment of human ATG13 (AA: 339-550) expressed in E. Coli. | |
| Primary | |
| 56.6 kDa |
| 6B2E12 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| autophagy-related protein 13, autophagy related 13, KIAA0652 | |
| ATG13 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title