missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG13 Antibody (6B2E12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52418
This item is not returnable.
View return policy
Description
ATG13 Monoclonal specifically detects ATG13 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| ATG13 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| autophagy-related protein 13, autophagy related 13, KIAA0652 | |
| Mouse | |
| 56.6 kDa | |
| 0.1 mg | |
| Phospho Specific | |
| 9776 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| 6B2E12 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ATG13 | |
| Purified recombinant fragment of human ATG13 (AA: 339-550) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction