missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG12 Antibody (2F6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | ATG12 |
|---|---|
| Clone | 2F6 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30038862
|
Novus Biologicals
NBP3-26495-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30033497
|
Novus Biologicals
NBP3-26495-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG12 Monoclonal antibody specifically detects ATG12 in Human samples. It is validated for ELISASpecifications
| ATG12 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog | |
| A synthesized peptide derived from Human ATG12 [UniProt O94817] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 2F6 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 9140 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title