missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG12 Antibody (2F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26495-100ul
This item is not returnable.
View return policy
Description
ATG12 Monoclonal antibody specifically detects ATG12 in Human samples. It is validated for ELISA
Specifications
| ATG12 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2F6 | |
| ELISA | |
| Apg12 (autophagy 12, S. cerevisiae)-like, APG12 autophagy 12-like (S. cerevisiae), APG12HAPG12, APG12-like, ATG12 autophagy related 12 homolog (S. cerevisiae), Autophagy-related protein 12, FBR93, ubiquitin-like protein ATG12, yeast) homolog | |
| A synthesized peptide derived from Human ATG12 [UniProt O94817] | |
| 100 μL | |
| Apoptosis, Autophagy, Neuroscience | |
| 9140 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction