missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG10 Recombinant Rabbit Monoclonal Antibody (23GB4860), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ATG10 |
|---|---|
| Clone | 23GB4860 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089147
![]() |
Invitrogen™
MA552752 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089139
|
Invitrogen™
MA552753 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG10 Recombinant Monoclonal antibody specifically detects ATG10 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ATG10 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9H0Y0 | |
| 83734 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4860 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5330424L23Rik; 5430428K15Rik; Agp10; AI852123; APG10; APG10L; APG10-like; Apg10p; ATG10; ATG10 autophagy related 10 homolog; Atg10l; autophagy related 10; autophagy related 10 homolog; autophagy-related 10; autophagy-related protein 10; DKFZp586I0418; FLJ13954; mAPG10; pp12616; Ubiquitin-like-conjugating enzyme ATG10 | |
| A synthesized peptide derived from human Apg10 (Atg10) (170-220AA). | |
| Primary | |
| -20°C | |
| ATG10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title