missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATG10 Recombinant Rabbit Monoclonal Antibody (23GB4860), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552752
This item is not returnable.
View return policy
Description
ATG10 is involved in autophagy which is a process for the bulk degradation of cytosolic compartments by lysosomes/vacuoles. The formation of autophagosomes involves a dynamic rearrangement of the membrane for which two ubiquitin-like modifications (the conjugation of APG12p and the modification of a soluble form of MAP-LC3 to a membrane-bound form) are essential. APG10 is an E2-like enzyme involved in two ubiquitin-like modifications essential for autophagosome formation, ATG12 -ATG5 conjugation and modification of a soluble form of MAP-LC3 (MAP1LC3A), a homolog of yeast Apg8, to a membrane-bound form. The protein ATG10 is ubiquitously expressed and the human APG10L gene is localized on chromosome 5q14.2.
Specifications
| ATG10 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATG10 | |
| 5330424L23Rik; 5430428K15Rik; Agp10; AI852123; APG10; APG10L; APG10-like; Apg10p; ATG10; ATG10 autophagy related 10 homolog; Atg10l; autophagy related 10; autophagy related 10 homolog; autophagy-related 10; autophagy-related protein 10; DKFZp586I0418; FLJ13954; mAPG10; pp12616; Ubiquitin-like-conjugating enzyme ATG10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 83734 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4860 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9H0Y0 | |
| ATG10 | |
| A synthesized peptide derived from human Apg10 (Atg10) (170-220AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction