missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Rabbit anti-Human, Clone: S05-4E7, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | ATF6 |
|---|---|
| Clone | S05-4E7 |
| Dilution | Western Blot 1:100-1:2000, ELISA, Immunohistochemistry |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18333144
|
Novus Biologicals
NBP3-14903-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309421
|
Novus Biologicals
NBP3-14903-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATF6 Monoclonal antibody specifically detects ATF6 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| ATF6 | |
| Western Blot 1:100-1:2000, ELISA, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| Recombinant protein of human ATF6 (Uniprot#: P18850) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S05-4E7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 22926 | |
| IgG | |
| Affinity purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title