missing translation for 'onlineSavingsMsg'
Learn More

ATF6 Rabbit anti-Human, Clone: S05-4E7, Novus Biologicals™

Product Code. 18333144 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333144 25 μg 25µL
18309421 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18333144 Supplier Novus Biologicals Supplier No. NBP31490325UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ATF6 Monoclonal antibody specifically detects ATF6 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATF6
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone S05-4E7
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA, Immunohistochemistry
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha
Host Species Rabbit
Immunogen Recombinant protein of human ATF6 (Uniprot#: P18850)
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR)
Primary or Secondary Primary
Gene ID (Entrez) 22926
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.