missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF2, p Thr69 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£219.00 - £340.00
Specifications
| Antigen | ATF2 (p Thr69) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263875
|
Novus Biologicals
NB100-81798-0.025ML |
0.025 mL |
£219.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260260
|
Novus Biologicals
NB100-81798 |
0.05 mL |
£340.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATF2 Polyclonal specifically detects ATF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ATF2 (p Thr69) | |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
| ATF2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Phospho Specific | |
| P15336 | |
| 1386 | |
| IgG | |
| Affinity Purified | |
| 70 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title