missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF2, p Thr69 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81798
This item is not returnable.
View return policy
Description
ATF2 Polyclonal specifically detects ATF2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ATF2 (p Thr69) | |
| Polyclonal | |
| Unconjugated | |
| PBS (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, 50% glycerol with 0.02% Sodium Azide | |
| activating transcription factor 2formerly, cAMP responsive element binding protein 2, cAMP-responsive element-binding protein 2, CREBP1, cyclic AMP-dependent transcription factor ATF-2, MGC111558 | |
| Rabbit | |
| 70 kDa | |
| 0.05 mL | |
| Phospho Specific | |
| 1386 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:500, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| P15336 | |
| ATF2 | |
| The antiserum was produced against synthesized phosphopeptide derived from human ATF2 around the phosphorylation site of threonine 69 or 51 (D-Q-TP-P-T). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of ATF2 only when phosphorylated at threonine 69 or 51. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction