missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ataxin 1 Antibody (S76-8), Novus Biologicals™
Mouse Monoclonal Antibody
£186.00 - £427.00
Specifications
| Antigen | Ataxin 1 |
|---|---|
| Clone | S76-8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664816
|
Novus Biologicals
NBP2-42186-0.025mg |
0.025 mg |
£186.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666556
|
Novus Biologicals
NBP2-42186 |
0.1 mg |
£427.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ataxin 1 Monoclonal antibody specifically detects Ataxin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Ataxin 1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P54254 | |
| 6310 | |
| IgG2b | |
| Protein G purified |
| S76-8 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 50% Glycerol | |
| ataxin 1, ataxin 1), ataxin-1, ATX1spinocerebellar ataxia 1 (olivopontocerebellar ataxia 1, autosomal dominant, SCA1D6S504E, Spinocerebellar ataxia type 1 protein | |
| Synthetic peptide amino acids 164-197 (ATTPSQRSQLEAYSTLLANMGSLSQAPGHKVEPP) of mouse Ataxin-1. Rat: 100% identity (34/34 amino acids identical). Human: 88% identity (30/34 amino acids identical). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title