missing translation for 'onlineSavingsMsg'
Learn More

Astryl, TRC

Product Code. 30396118
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396118 2 mg 2mg
30397699 10 mg 10mg
30408686 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396118 Supplier Toronto Research Chemicals Supplier No. TRCH9545752MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-87-6
Molecular Formula C8H10NO5As
Molecular Weight (g/mol) 275.09
Synonym Glycarsamide, Glycarsiamidon, N-Glycoloylarsanilic Acid, NSC 163966, NSC 221701, [4-(2-Hydroxyacetamido)phenyl]arsonic Acid, p-(Glycoloylamino)benzenearsonic Acid, p-(Glycoloylamino)phenylarsonic Acid, p-(Hydroxyacetamido)phenylarsonic Acid
IUPAC Name [4-[(2-hydroxyacetyl)amino]phenyl]arsonic acid
SMILES OCC(=O)Nc1ccc(cc1)[As](=O)(O)O

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C8H10AsNO5/c11-5-8(12)10-7-3-1-6(2-4-7)9(13,14)15/h1-4,11H,5H2,(H,10,12)(H2,13,14,15)
Formula Weight 274.9775
Chemical Name or Material Astryl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.