CAS RN 144-87-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Astryl, TRC
CAS: 144-87-6 Molecular Formula: C8H10NO5As Molecular Weight (g/mol): 275.09 Synonym: Glycarsamide,Glycarsiamidon,N-Glycoloylarsanilic Acid,NSC 163966,NSC 221701,[4-(2-Hydroxyacetamido)phenyl]arsonic Acid,p-(Glycoloylamino)benzenearsonic Acid,p-(Glycoloylamino)phenylarsonic Acid,p-(Hydroxyacetamido)phenylarsonic Acid IUPAC Name: [4-[(2-hydroxyacetyl)amino]phenyl]arsonic acid SMILES: OCC(=O)Nc1ccc(cc1)[As](=O)(O)O