Learn More
DiscoverX AssayComplete™ Cell Plating 36 Reagent
Product Code. 17323439
Encompass
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. 17323439
Brand: DiscoverX 930563R36B
AssayComplete™ Cell Plating 36 Reagent is a reagent used to plate and assay DiscoverX cell lines. This product is provided as a bottle of 500mL.
AssayComplete™ Cell Plating 36 Reagent is an optimized cell plating reagent for use in plating of DiscoverX cell lines for assays. This reagent was optimized for maximal cell health and ligand responsiveness during assays while eliminating hormones and growth components which may lead to increased basal activity. This product is provided as a bottle of 500mL. Please refer to cell line specific datasheet to determine the recommended cell plating reagent.
Specifications
For Use With (Application) Cell Plating Reagents
Quantity 500 mL