Learn More
DiscoverX AssayComplete™ Cell Plating 15 Reagent
Product Code. 17323409
Encompass
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 17323409
Brand: DiscoverX 930563R15A
AssayComplete™ Cell Plating 15 Reagent is a reagent used to plate and assay DiscoverX cell lines. This product is provided as a bottle of 100mL.
AssayComplete™ Cell Plating 15 Reagent is an optimized cell plating reagent for use in plating of DiscoverX cell lines for assays. This reagent was optimized for maximal cell health and ligand responsiveness during assays while eliminating hormones and growth components which may lead to increased basal activity. This product is provided as a bottle of 100mL. Please refer to cell line specific datasheet to determine the recommended cell plating reagent.
Specifications
For Use With (Application) Cell Plating Reagents
Quantity 100 mL