missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate beta hydroxylase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69229
This item is not returnable.
View return policy
Description
Aspartate beta hydroxylase Polyclonal specifically detects Aspartate beta hydroxylase in Human samples. It is validated for Western Blot.
Specifications
| Aspartate beta hydroxylase | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ASPH | |
| Synthetic peptides corresponding to ASPH(aspartate beta-hydroxylase) The peptide sequence was selected from the N terminal of ASPH. Peptide sequence MAEDKETKHGGHKNGRKGGLSGTSFFTWFMVIALLGVWTSVAVVWFDLVD. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Porcine: 86%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| AAH, ASP beta-hydroxylase, aspartate beta-hydroxylaseA beta H-J-J, aspartyl/asparaginyl-beta-hydroxylase, BAH, cardiac junctin, CASQ2BP1, EC 1.14.11.16, HAAHaspartyl/asparaginyl beta-hydroxylase, humbug, JCTN, junctate, junctin, Peptide-aspartate beta-dioxygenase | |
| Rabbit | |
| 25 kDa | |
| 100 μL | |
| Cancer | |
| 444 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction