missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASL Monoclonal Antibody (24GB6585)

Mouse Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | ASL |
|---|---|
| Clone | 24GB6585 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275867
![]() |
Invitrogen™
MA557341 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276604
|
Invitrogen™
MA557342 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASL Monoclonal Antibody for Western Blot
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| ASL | |
| -20°C | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| ASL | |
| 2510006M18Rik; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; Asl | |
| ASL | |
| Primary | |
| Affinity chromatography |
| 24GB6585 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04424 | |
| 435 | |
| Recombinant protein of human ASL (aa 1-464). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title