missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASL Monoclonal Antibody (24GB6585)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557342
This item is not returnable.
View return policy
Description
ASL Monoclonal Antibody for Western Blot
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| ASL | |
| Monoclonal | |
| Unconjugated | |
| ASL | |
| 2510006M18Rik; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; Asl | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 435 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6585 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04424 | |
| ASL | |
| Recombinant protein of human ASL (aa 1-464). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido