missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £234.00
Specifications
| Antigen | ASK1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18030396
|
Novus Biologicals
NB100-56077SS |
0.025 mL |
£157.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008395
|
Novus Biologicals
NB100-56077 |
0.05 mL |
£234.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASK1 Polyclonal specifically detects ASK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| ASK1 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| Whole antisera with 0.02% Sodium Azide | |
| 4217 | |
| A synthetic peptide corresponding to amino acids 1198 to 1216(CHEEQPSNQTVRRPQAVIED) of human Ask1 was used as immunogen; GenBank no. NP_005914.1. Human Ask1 is a 1374 amino acid protein. The peptide immunogen sequence is 100% conserved in Ask1 variant (BAD92330.1), a 1455 amino acid protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5 | |
| MAP3K5 | |
| IgG | |
| Unpurified | |
| ASK1 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title