missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56077
This item is not returnable.
View return policy
Description
ASK1 Polyclonal specifically detects ASK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| ASK1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Apoptosis signal-regulating kinase 1, ASK-1, ASK1MEKK 5, EC 2.7.11, MAP/ERK kinase kinase 5, MAPK/ERK kinase kinase 5, MAPKKK5EC 2.7.11.25, MEK kinase 5, MEKK5apoptosis signal regulating kinase 1, mitogen-activated protein kinase kinase kinase 5 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| ASK1 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| MAP3K5 | |
| A synthetic peptide corresponding to amino acids 1198 to 1216(CHEEQPSNQTVRRPQAVIED) of human Ask1 was used as immunogen; GenBank no. NP_005914.1. Human Ask1 is a 1374 amino acid protein. The peptide immunogen sequence is 100% conserved in Ask1 variant (BAD92330.1), a 1455 amino acid protein. | |
| 0.05 mL | |
| Apoptosis | |
| 4217 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction