missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASH2L Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£176.00 - £318.00
Specifications
| Antigen | ASH2L |
|---|---|
| Concentration | 2 mg/ml |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18302615
|
Novus Biologicals
NBP3-18668-25UL |
25 μg |
£176.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18306205
|
Novus Biologicals
NBP3-18668-50UG |
50 μg |
£318.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Human samples. It is validated for Western BlotSpecifications
| ASH2L | |
| Western Blot 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q9UBL3 | |
| 9070 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| 2 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Transcription Factors and Regulators | |
| PBS | |
| ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
| Polyclonal antibody raised in rabbit against human ASH2L (absent, small, or homeotic 2), using a recombinant protein. | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title