missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASH2L Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18668-50UG
This item is not returnable.
View return policy
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Human samples. It is validated for Western Blot
Specifications
| ASH2L | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2 | |
| Polyclonal antibody raised in rabbit against human ASH2L (absent, small, or homeotic 2), using a recombinant protein. | |
| 50 μg | |
| DNA Repair, Transcription Factors and Regulators | |
| 9070 | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 2 mg/ml | |
| Western Blot 1:1000 | |
| Q9UBL3 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction