missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASGR1 Mouse anti-Human, Mouse, Porcine, Clone: 1G5F5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ASGR1 |
|---|---|
| Clone | 1G5F5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808815
|
Proteintech
66692-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898805
|
Proteintech
66692-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The asialoglycooprotein receptor (ASGPR) is a transmembrane glycoprotein (42 kDa) which mediates binding, internalisation and degradation of extracellular glycoproteins that have exposed terminal galactose residues. The receptor is expressed on the surface of the hepatocytes in a polar manner, i.e. it is present on the sinusoidal and lateral palasma membranes but not on the bile canalicular membrane. The mammalian hepatic ASGPR mediate the endocytosis and degradation of serum proteins from which terminal sialic residues have been removed.The rat asialoglycoprotein receptor (ASGPR) consists of three polypeptide subunits: rat hepatic lectin (RHL)-1, RHL-2 and RHL-3. The monoclonal antibody 8D7 recognizes a subunit-specific epitope of RHL-1 of rat ASGPR. The monoclonal antibody 8D7 is cross reactive with human ASGPR.Specifications
| ASGR1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| ASGP R 1, ASGPR, ASGPR 1, ASGR1, asialoglycoprotein receptor 1, CLEC4H1, Hepatic lectin H1, HL 1, Hs.12056 | |
| ASGR1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1G5F5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Pig, Mouse | |
| P07306, P34927 | |
| 100625649, 11889, 432 | |
| ASGR1 Fusion Protein Ag2310 | |
| Primary | |
| -20°C | |
| ASGR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title