missing translation for 'onlineSavingsMsg'
Learn More

ASCL1/Mash1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£183.00 - £255.00

Specifications

Antigen ASCL1/Mash1
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-5 uL/mg of lysate
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655507
View Documents
Novus Biologicals
NBP2-99418-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18672259
View Documents
Novus Biologicals
NBP2-99418-50ul
50 μg
£183.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ASCL1/Mash1 Polyclonal antibody specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications

Specifications

ASCL1/Mash1
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Neuroscience
PBS (pH 7.0)
429
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunoprecipitation 1-5 uL/mg of lysate
Polyclonal
Purified
RUO
Mouse
achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, hASH1, HASH1achaete scute protein
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ASCL1/Mash1
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.