missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCL1/Mash1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99418-100ul
This item is not returnable.
View return policy
Description
ASCL1/Mash1 Polyclonal antibody specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ASCL1/Mash1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 uL/mg of lysate | |
| achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, hASH1, HASH1achaete scute protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ASCL1/Mash1 | |
| 100 μg | |
| Neuroscience | |
| 429 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction