Learn More
ASC/TMS1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10500-1-AP-150UL
Description
The ASC/TMS1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ASC/TMS1. This antibody recognizes human antigen. The ASC/TMS1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| ASC/TMS1 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 29108 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human |