missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56645
This item is not returnable.
View return policy
Description
ASB3 Polyclonal specifically detects ASB3 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot.
Specifications
| ASB3 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| Q9Y575 | |
| ASB3 | |
| This antibody was developed against synthetic peptides corresponding to amino acids 63-75 and 166-180 of human Asb-3 (NP_665862). | |
| Unpurified | |
| RUO | |
| Primary | |
| Cross reacts with Human. It does not cross-react with Mouse or Rat Asb-3 . | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Whole antisera with PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide | |
| ankyrin repeat and SOCS box containing 3, ankyrin repeat and SOCS box protein 3, ankyrin repeat and SOCS box-containing 3, ASB-3MGC132002, FLJ10123, FLJ10421, MGC12531, MGC996 | |
| Rabbit | |
| 57.745 kDa | |
| 0.2 mL | |
| Signal Transduction | |
| 51130 | |
| Human, Mouse (Negative), Rat (Negative) | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction