missing translation for 'onlineSavingsMsg'
Learn More

ASB3 Antibody, Novus Biologicals™

Product Code. p-5061882 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
Unit Size:
0.05mL
0.2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18077163 0.05 mL 0.05mL
18055704 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18077163 Supplier Novus Biologicals Supplier No. NB10056645SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ASB3 Polyclonal specifically detects ASB3 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation Whole antisera with PBS and 0.1 mg/ml BSA with 0.02% Sodium Azide
Gene Accession No. Q9Y575
Gene Alias ankyrin repeat and SOCS box containing 3, ankyrin repeat and SOCS box protein 3, ankyrin repeat and SOCS box-containing 3, ASB-3MGC132002, FLJ10123, FLJ10421, MGC12531, MGC996
Gene Symbols ASB3
Host Species Rabbit
Immunogen This antibody was developed against synthetic peptides corresponding to amino acids 63-75 and 166-180 of human Asb-3 (NP_665862).
Molecular Weight of Antigen 57.745 kDa
Purification Method Unpurified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51130
Test Specificity Cross reacts with Human. It does not cross-react with Mouse or Rat Asb-3 .
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.