missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11274-1-AP-20UL
This item is not returnable.
View return policy
Description
Sphingolipids are hydrolyzed by ceramidases to yield sphingosine and fatty acids. These ceramidases are classified according to the pH range that supports their optimal activity. ASAH1 is an acid ceramidase and key regulator of ceramide metabolism. Mutations in this gene results in Farber Lipogranulomatosis, a fatal human genetic disorder that results in the painful swelling of the joints and tendons and pulmonary insufficiency, while a complete knockout of its expression is lethal in mice. Recent studies have shown elevated levels of ASAH1 in Alzheimer′s disease (AD) patients correlating with a reduction in sphingomyelin and elevation of ceramide. Pretreatment of cultured neurons with recombit AHAH1 prevented the cells from undergoing A-beta (Ab)-induced apoptosis. Multiple isoforms of this protein are known to exist.Specifications
| ASAH1 | |
| Polyclonal | |
| Unconjugated | |
| ASAH1 | |
| AC, Acid ceramidase, Acylsphingosine deacylase, ASAH, ASAH1, HSD 33, PHP, PHP32, Putative 32 kDa heart protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11886, 427, 84431 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13510, Q6P7S1, Q9WV54 | |
| Asah1 | |
| ASAH1 Fusion Protein Ag1799 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction