missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ASAH1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £395.00

Specifications

Antigen ASAH1
Dilution Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067082
View Documents
Novus Biologicals™
NBP3-30951-100ug
100 μg
£395.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067144
View Documents
Novus Biologicals™
NBP3-30951-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ASAH1 Polyclonal antibody specifically detects ASAH1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

ASAH1
ELISA, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Mouse, Rat
ACDase, ACEC 3.5.1.23, Acid CDase, acid ceramidase, Acylsphingosine deacylase, ASAH, FLJ21558, FLJ22079, N-acylsphingosine amidohydrolase, N-acylsphingosine amidohydrolase (acid ceramidase) 1, PHP, PHP32N-acylsphingosine amidohydrolase (acid ceramidase), Putative 32 kDa heart protein
Synthetic peptide derived from the C terminal region of the ASAH1 protein.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL
Polyclonal
Purified
RUO
Phosphate buffered saline
427
IgG
Antigen Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.