missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ASAH1 Polyclonal antibody specifically detects ASAH1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ASAH1 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot : 1-5 μg/mL, ELISA, Immunohistochemistry-Paraffin : 10 μg/mL |
| Formulation | Phosphate buffered saline |
| Gene Alias | ACDase, ACEC 3.5.1.23, Acid CDase, acid ceramidase, Acylsphingosine deacylase, ASAH, FLJ21558, FLJ22079, N-acylsphingosine amidohydrolase, N-acylsphingosine amidohydrolase (acid ceramidase) 1, PHP, PHP32N-acylsphingosine amidohydrolase (acid ceramidase), Putative 32 kDa heart protein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the C terminal region of the ASAH1 protein. |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?