missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79795
This item is not returnable.
View return policy
Description
ART1 Polyclonal specifically detects ART1 in Human samples. It is validated for Western Blot.
Specifications
| ART1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 86%; Dog: 85%; Mouse: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P52961 | |
| ART1 | |
| Synthetic peptide directed towards the C terminal of human ART1The immunogen for this antibody is ART1 (NP_004305). Peptide Sequence: KHSTYNCEYIKDKKCKSGPCHLDNSAMGQSPLSAVWSLLLLLWFLVVRAF | |
| Affinity purified | |
| RUO | |
| 417 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction