missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
£177.00 - £459.00
Specifications
| Antigen | ARP10 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18464182
|
Novus Biologicals
NBP1-91682-25ul |
25 μL |
£177.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707293
|
Novus Biologicals
NBP1-91682 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARP10 Polyclonal specifically detects ARP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ARP10 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q6NTF7 | |
| 164668 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CSSCAWELVDFIKAHDHLNLGIFASRLYYHWCKPQQKGLRLLCGSQVPVEVMGFPEFADCWENFVDHEKPLSFNPYKMLEELDKNSRAIKRRLERIKQS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 24 kDa |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| APOBEC-related protein 10, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H, ARP-10, ARP10Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H, DNA dC->dU-editing enzyme APOBEC-3H, EC 3.5.4.- | |
| APOBEC3H | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto