Learn More
ARP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-91682
This item is not returnable.
View return policy
Description
ARP10 Polyclonal specifically detects ARP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| APOBEC-related protein 10, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H, ARP-10, ARP10Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H, DNA dC->dU-editing enzyme APOBEC-3H, EC 3.5.4.- | |
| Affinity Purified | |
| RUO | |
| IgG |
| Rabbit | |
| 0.1 mL | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |