missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aromatase Antibody (H4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£169.00 - £397.00
Specifications
| Antigen | Aromatase |
|---|---|
| Clone | H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:50-1:5000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230163
|
Novus Biologicals
NBP2-61939 |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678166
|
Novus Biologicals
NBP2-61939-0.025mg |
0.025 mg |
£169.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
| Aromatase | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Rat, Pig | |
| ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
| CYP19A1 | |
| IgG2a | |
| Protein A purified |
| H4 | |
| Western Blot 1:50-1:5000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 1588 | |
| Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK) | |
| Primary | |
| 55 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title