missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aromatase Antibody (H4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61939-0.025mg
This item is not returnable.
View return policy
Description
Aromatase Monoclonal antibody specifically detects Aromatase in Human, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
| Aromatase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:50-1:5000, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 | |
| ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
| Mouse | |
| 55 kDa | |
| 0.025 mg | |
| Primary | |
| Human, Rat, Pig | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| H4 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CYP19A1 | |
| Synthetic peptide corresponding to AAs 376-390 of human aromatase (KALEDDVIGYPVKK) | |
| Protein A purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience | |
| 1588 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction