missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL8B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56544
This item is not returnable.
View return policy
Description
ARL8B Polyclonal specifically detects ARL8B in Human samples. It is validated for Western Blot.
Specifications
| ARL8B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ADP-ribosylation factor-like 10C, ADP-ribosylation factor-like 8B, ADP-ribosylation factor-like protein 10C, ADP-ribosylation factor-like protein 8B, ARL10C, FLJ10702, Gie1, Novel small G protein indispensable for equal chromosome segregation 1 | |
| Rabbit | |
| 22 kDa | |
| 100 μL | |
| Signal Transduction | |
| 55207 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NVJ2 | |
| ARL8B | |
| Synthetic peptides corresponding to ARL8B(ADP-ribosylation factor-like 8B) The peptide sequence was selected from the middle region of ARL8B (NP_060654). Peptide sequence DIGGQPRFRSMWERYCRGVNAIVYMIDAADREKIEASRNELHNLLDKPQL. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 100%; Bovine: 100%; Chicken: 100%; Canine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 92%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction