missing translation for 'onlineSavingsMsg'
Learn More

ARL13B Guinea Pig Polyclonal Antibody, Proteintech

Pig Polyclonal Antibody

£119.00 - £295.00

Specifications

Antigen ARL13B
Concentration 750 μg/mL
Applications Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234583
View Documents
Proteintech
30332-1-AP-150UL
150 μL
ONLINE EXCLUSIVE PRICE
£349.00
£295.00 / 150µL
Save £54.00
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235402
View Documents
Proteintech
30332-1-AP-20UL
20 μL
ONLINE EXCLUSIVE PRICE
£135.00
£119.00 / 20µL
Save £16.00
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ARL13B antibody from Proteintech is a guinea pig polyclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human antigen. The ARL13B antibody has been validated for the following applications: WB, IF/ICC, IP, ELISA analysis.
Specifications

Specifications

ARL13B
Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA
Unconjugated
Guinea Pig
Human
ARL2L1; DKFZp686E2075; DKFZp686L2472; DKFZp686M2074; DKFZp761H079; JBTS8; MGC120611; MGC120612
IgG
Antigen affinity purification
Polyclonal Antibody
750 μg/mL
Polyclonal
Liquid
RUO
200894
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.