missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Guinea Pig Polyclonal Antibody, Proteintech
Pig Polyclonal Antibody
Brand: Proteintech 30332-1-AP-20UL
This item is not returnable.
View return policy
Description
The ARL13B antibody from Proteintech is a guinea pig polyclonal antibody generated with recombinant protein of human ARL13B. This antibody recognizes human antigen. The ARL13B antibody has been validated for the following applications: WB, IF/ICC, IP, ELISA analysis.Specifications
| ARL13B | |
| Polyclonal | |
| Unconjugated | |
| Guinea Pig | |
| Antigen affinity purification | |
| RUO | |
| 200894 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 750 μg/mL | |
| ARL2L1; DKFZp686E2075; DKFZp686L2472; DKFZp686M2074; DKFZp761H079; JBTS8; MGC120611; MGC120612 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 48-55 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction