missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARK5/NUAK1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£177.00 - £366.00
Specifications
| Antigen | ARK5/NUAK1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697651
|
Novus Biologicals
NBP2-92357-0.02ml |
0.02 mL |
£177.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663440
|
Novus Biologicals
NBP2-92357-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARK5/NUAK1 Polyclonal antibody specifically detects ARK5/NUAK1 in Human, Rat samples. It is validated for Western BlotSpecifications
| ARK5/NUAK1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 50% glycerol, pH7.3. | |
| 9891 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| AMPK-related protein kinase 5, ARK5AMP-activated protein kinase family member 5, EC 2.7.11, EC 2.7.11.1, KIAA0537NUAK family SNF1-like kinase 1, NUAK family, SNF1-like kinase, 1, NuaK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 362-661 of human NUAK1 (NP_055655.1). LERQRSLKKSKKENDFAQSGQDAVPESPSKLSSKRPKGILKKRSNSEHRSHSTGFIEGVVGPALPSTFKMEQDLCRTGVLLPSSPEAEVPGKLSPKQSATMPKKGILKKTQQRESGYYSSPERSESSELLDSNDVMGSSIPSPSPPDPARVTSHSLSCRRKGILKHSSKYSAGTMDPALVSPEMPTLESLSEPGVPAEGLSRSYSRPSSVISDDSVLSSDSFDLLDLQENRPARQRIRSCVSAENFLQIQDFEGLQNRPRPQYLKRYRNRLADSSFSLLTDMDDVTQVYKQALEICSKLN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title