missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARK5/NUAK1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92357-0.1ml
This item is not returnable.
View return policy
Description
ARK5/NUAK1 Polyclonal antibody specifically detects ARK5/NUAK1 in Human, Rat samples. It is validated for Western Blot
Specifications
| ARK5/NUAK1 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| AMPK-related protein kinase 5, ARK5AMP-activated protein kinase family member 5, EC 2.7.11, EC 2.7.11.1, KIAA0537NUAK family SNF1-like kinase 1, NUAK family, SNF1-like kinase, 1, NuaK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 362-661 of human NUAK1 (NP_055655.1). LERQRSLKKSKKENDFAQSGQDAVPESPSKLSSKRPKGILKKRSNSEHRSHSTGFIEGVVGPALPSTFKMEQDLCRTGVLLPSSPEAEVPGKLSPKQSATMPKKGILKKTQQRESGYYSSPERSESSELLDSNDVMGSSIPSPSPPDPARVTSHSLSCRRKGILKHSSKYSAGTMDPALVSPEMPTLESLSEPGVPAEGLSRSYSRPSSVISDDSVLSSDSFDLLDLQENRPARQRIRSCVSAENFLQIQDFEGLQNRPRPQYLKRYRNRLADSSFSLLTDMDDVTQVYKQALEICSKLN | |
| 0.1 mL | |
| Protein Kinase | |
| 9891 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction