missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57038
This item is not returnable.
View return policy
Description
ARHGEF19 Polyclonal specifically detects ARHGEF19 in Human samples. It is validated for Western Blot.
Specifications
| ARHGEF19 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ephexin-2, FLJ33962, Rho guanine nucleotide exchange factor (GEF) 19, RP4-733M16.1, weakly similar to Rho GEF, WGEFrho guanine nucleotide exchange factor 19 | |
| Rabbit | |
| 89 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Bovine: 92%. | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8IW93 | |
| ARHGEF19 | |
| Synthetic peptides corresponding to ARHGEF19 (Rho guanine nucleotide exchange factor (GEF) 19) The peptide sequence was selected from the middle region of ARHGEF19 (NP_694945). Peptide sequence: RFLLNSVLYQEYSDVASARELRRQQREEEGPGDEAEGAEEGPGPPRANLS. | |
| Affinity purified | |
| RUO | |
| 128272 | |
| Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction