missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ARHGAP5 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856645
|
Proteintech
55165-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846645
|
Proteintech
55165-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to ARHGAP5.
The 34-amino acid tetratricopeptide repeat (TPR) motif is found in a variety of proteins and may mediate protein-protein or protein-membrane interactions.Specifications
| ARHGAP5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| P97393, Q13017 | |
| 11855, 299012, 394 | |
| Peptide | |
| Primary | |
| -20°C | |
| ARHGAP5 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ARHGAP5, GFI2, p190 B, RhoGAP5 | |
| ARHGAP5 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title